missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human DYRK1A, GST Tag Recombinant Protein
DYRK1A Recombinant Human Protein
Brand: Invitrogen™ PV3997
Additional Details : Weight : 0.00001kg
Description
DYRK1A is a dual specificity kinase involved in cell proliferation. The gene for DYRK1A is located in the Down syndrome critical region of chromosome 21, and has been implicated in the pathogenesis of this disease.Specifications
NP_001387 | |
Protein Assays and Analysis, Kinase Assay | |
113kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
DYRK1A, DYRK1 | |
DYRK1A was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL DYRKtide peptide substrate (RRRFRPASPLRGPPK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
DYRK1A Recombinant Human Protein | |
Human | |
Baculovirus | |
85% by SDS-PAGE |
0.16mg/mL | |
Frozen | |
DYRK1A Recombinant Human Protein | |
–80°C | |
1 Each | |
DYRK1A Recombinant Human Protein | |
DYRK | |
2880nmoles of phosphate transferred to DYRKtide peptide substrate (RRRFRPASPLRGPPK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.83μg/mL | |
GST Tag | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.