missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DYRK1B, GST Tag Recombinant Protein
DYRK1B Recombinant Human Protein
Brand: Invitrogen PV4649
Additional Details : Weight : 0.00900kg
Description
DYRK1B is a member of the DYRK family of protein kinases and is proposed to be involved in nuclear functions.Specifications
NP_004705 | |
9149 | |
10 μg | |
DYRK Kinase Family, DYRKs | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100 Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMCG Group | |
Z'-LYTE™ | |
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1B | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
96.7 kDa | |
DYRK1B, MIRK | |
Human | |
GST Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
DYRK1B | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |