Learn More
Invitrogen™ DYRK3 Recombinant Human Protein
DYRK3 belongs to the DYRK family of dual-specificity protein kinases.
Brand: Invitrogen™ PV3998
Additional Details : Weight : 0.00900kg
Description
DYRK3 catalyzes phosphorylation of serine/threonine and tyrosine residues. The members of the DYRK family share structural similarity, but differ in their substrate specificity, suggesting their involvement in different cellular functions.
Specifications
NP_003573 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
DYRK3 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
93.2kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
DYRK Kinase Family | |
Recombinant | |
Baculovirus |