missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYRK4 Recombinant Human Protein
DYRK4 Recombinant Human Protein
Brand: Invitrogen PV3871
Additional Details : Weight : 0.05250kg
Description
Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development.Specifications
NP_003836.1 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
88.8kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
DYRK4 | |
DYRK4 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL DYRKtide peptide substrate (RRRFRPASPLRGPPK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
DYRK4 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.47mg/mL | |
114294, 8798 | |
DYRK4 Recombinant Human Protein | |
–80°C | |
1 Each | |
DYRK4 Recombinant Human Protein | |
DYRK | |
146nmoles of phosphate transferred to DYRKtide peptide substrate (RRRFRPASPLRGPPK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
GST Tag | |
Kinase and inhibitors | |
55% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.