Läs mer
(E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone Hydrochloride, TRC
Produktkod. 30465232
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465232 100 mg
30478115 500 mg
30476791 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30465232 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM725995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 938071-78-4
Molekylformel C17H24N2O . x(HCl)
Molekylvikt (g/mol) 308.8
Synonym Membrane Traffic Inhibitor, A5 Hydrochloride
IUPAC-namn (E)-1-(4-(4-(2-methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethan-1-one hydrochloride
LEDER CC(C1=CC=C(N2CCN(C/C(C)=C/C)CC2)C=C1)=O.Cl
Specifikationer
Kemiskt namn eller material (E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C17H24N2O.ClH/c1-4-14(2)13-18-9-11-19(12-10-18)17-7-5-16(6-8-17)15(3)20;/h4-8H,9-13H2,1-3H3;1H/b14-4+;