Läs mer
(E)-11,13-Tetradecadienyl Acetate, TRC
Produktkod. 30428427
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30428427 10 mg
1 options
Produktkod. 30428427 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29175510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-11,13-Tetradecadienyl Acetate
CAS 80625-43-0
Molekylformel C16 H28 O2
Kvantitet 10 mg
Synonym trans-11,13-Tetradecadienyl Acetate, (E)-11,13-Tetradecadien-1-ol Acetate
InChI formel InChI=1S/C16H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h3-5H,1,6-15H2,2H3/b5-4+
LEDER CC(=O)OCCCCCCCCCC\C=C\C=C
IUPAC-namn [(11E)-tetradeca-11,13-dienyl] acetate
Molekylvikt (g/mol) 252.392
Formel vikt 252.209