Läs mer
(E)-2-Chloro-3-imino-N,N-dimethylprop-1-en-1-amine Hydrochloride, TRC
Produktkod. 30132526
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132526 500 mg
30009956 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC371270500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1352804-27-3
Molekylformel C5 H9 Cl N2. H Cl
Molekylvikt (g/mol) 169.05
Synonym Etoricoxib Impurity 27;(E)-2-chloro-3-imino-N,N-dimethylprop-1-en-1-amine hydrochloride
IUPAC-namn (E)-2-chloro-3-imino-N,N-dimethyl-prop-1-en-1-amine;hydrochloride
LEDER Cl.CN(C)\C=C(\Cl)/C=N
Specifikationer
Kemiskt namn eller material (E)-2-chloro-3-imino-N,N-dimethyl-prop-1-en-1-amine hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C5H9ClN2.ClH/c1-8(2)4-5(6)3-7;/h3-4,7H,1-2H3;1H/b5-4+,7-3?;
Formel vikt 168.0221