Läs mer
E 215, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Sodium Ethyl Parahydroxybenzoate |
| CAS | 35285-68-8 |
| Molekylformel | C9 H9 O3 . Na |
| Kvantitet | 5 g |
| Synonym | Sodium Ethyl Parahydroxybenzoate, p-Hydroxybenzoic acid ethyl ester sodium deriv., 4-Hydroxybenzoic acid ethyl ester sodium salt, Ethyl 4-hydroxybenzoate sodium salt, Ethyl p-hydroxybenzoate sodium salt, Sodium ethyl p-hydroxybenzoate, Sodium p-(carbethoxy)phenoxide |
| InChI formel | InChI=1S/C9H10O3.Na/c1-2-12-9(11)7-3-5-8(10)6-4-7;/h3-6,10H,2H2,1H3;/q;+1/p-1 |
| LEDER | [Na+].CCOC(=O)c1ccc([O-])cc1 |
| IUPAC-namn | sodium;4-ethoxycarbonylphenolate |
| Molekylvikt (g/mol) | 188.16 |
| Formel vikt | 188.0449 |