Läs mer
(E)-3-(3-(Trifluoromethyl)phenyl)acrylic Acid, TRC
Produktkod. 30450715
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30450715 10 g
1 options
Produktkod. 30450715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77454610G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-3-(3-(Trifluoromethyl)phenyl)acrylic Acid
CAS 67801-07-4
Molekylformel C10H7F3O2
Kvantitet 10 g
Synonym (2E)-3-[3-(Trifluoromethyl)phenyl]-2-propenoic Acid, (E)-3-(3-Trifluoromethylphenyl)-2-propenoic Acid, (E)-3-(Trifluoromethyl)cinnamic Acid, (E)-m-(Trifluoromethyl)cinnamic Acid, trans-3-(3-Trifluoromethylphenyl)-2-propenoic Acid, trans-3-(3-Trifluoromethylphenyl)acrylic Acid, trans-3-(Trifluoromethyl)cinnamic Acid
InChI formel InChI=1S/C10H7F3O2/c11-10(12,13)8-3-1-2-7(6-8)4-5-9(14)15/h1-6H,(H,14,15)/b5-4+
LEDER OC(=O)\C=C\c1cccc(c1)C(F)(F)F
IUPAC-namn (E)-3-[3-(trifluoromethyl)phenyl]prop-2-enoic acid
Molekylvikt (g/mol) 216.16
Formel vikt 216.0398