Läs mer
(E)-(3-Cyano-2-methyl-2-propenyl)-phosphonic Acid Diethyl Ester (~10% Z-isomer), TRC
Produktkod. 30365480
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30365480 100 mg
1 options
Produktkod. 30365480 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC987498100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-(3-Cyano-2-methyl-2-propenyl)-phosphonic Acid Diethyl Ester
CAS 82648-70-2
Molekylformel C9H16NO3P
Kvantitet 100 mg
Synonym [(2E)-3-Cyano-2-methyl-2-propenyl]-phosphonic Acid Diethyl Ester
InChI formel InChI=1S/C9H16NO3P/c1-4-12-14(11,13-5-2)8-9(3)6-7-10/h6H,4-5,8H2,1-3H3/b9-6+
LEDER CCOP(=O)(C\C(=C\C#N)\C)OCC
IUPAC-namn (E)-4-diethoxyphosphoryl-3-methylbut-2-enenitrile
Molekylvikt (g/mol) 217.2