Läs mer
(E)-3-Methoxy-5-styrylphenol, TRC
Produktkod. 30472926
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30472926 2.5 mg 2.5mg
30472443 25 mg -
30481158 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2670032.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 35302-70-6
Molekylformel C15H14O2
Molekylvikt (g/mol) 226.27
Synonym Phenol, 3-methoxy-5-[(1E)-2-phenylethenyl]-, Phenol, 3-methoxy-5-(2-phenylethenyl)-, (E)-, (E)-3-Hydroxy-5-methoxystilbene, Pinosylvin 3-(methyl ether), Pinosylvin monomethyl ether, Pinosylvin, methyl ether
IUPAC-namn 3-methoxy-5-[(E)-2-phenylethenyl]phenol
LEDER COc1cc(O)cc(\C=C\c2ccccc2)c1
Specifikationer
Kemiskt namn eller material (E)-3-Methoxy-5-styrylphenol
Kvantitet 2.5 mg
InChI formel InChI=1S/C15H14O2/c1-17-15-10-13(9-14(16)11-15)8-7-12-5-3-2-4-6-12/h2-11,16H,1H3/b8-7+
Formel vikt 226.0994