Läs mer
(E)-4-Hydroxy Toremifene (~5% Z-isomer), TRC
Produktkod. 30137263
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137263 100 mg
30010745 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137263 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH954505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 177748-22-0
Molekylformel C26 H28 Cl N O2
Molekylvikt (g/mol) 421.96
Synonym (E)-4-[4-Chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]phenol, (E)-4-Hydroxytoremifene
IUPAC-namn 4-[(E)-4-chloro-1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-but-1-enyl]phenol
LEDER CN(C)CCOc1ccc(cc1)\C(=C(/CCCl)\c2ccccc2)\c3ccc(O)cc3
Specifikationer
Kemiskt namn eller material (E)-4-Hydroxy Toremifene
Kvantitet 100 mg
InChI formel InChI=1S/C26H28ClNO2/c1-28(2)18-19-30-24-14-10-22(11-15-24)26(21-8-12-23(29)13-9-21)25(16-17-27)20-6-4-3-5-7-20/h3-15,29H,16-19H2,1-2H3/b26-25+
Formel vikt 421.1809