Läs mer
(E)-4-Methyl-2-phenyl-2-pentenal (Stabilized with Alpha-tocopherol), TRC
Produktkod. 30464941
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464941 100 mg
30466281 500 mg
30471496 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464941 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM220353100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1474032-26-2
Molekylformel C12H14O
Molekylvikt (g/mol) 174.24
Synonym (alphaE)-alpha-(2-Methylpropylidene)benzeneacetaldehyde
IUPAC-namn (E)-4-methyl-2-phenylpent-2-enal
LEDER CC(C)C=C(C=O)c1ccccc1
Specifikationer
Kemiskt namn eller material (E)-4-Methyl-2-phenyl-2-pentenal
Kvantitet 100 mg
InChI formel InChI=1S/C12H14O/c1-10(2)8-12(9-13)11-6-4-3-5-7-11/h3-10H,1-2H3/b12-8-
Formel vikt 174.1045