Läs mer
3M™ E-A-R™ Classic™ SuperFit™ Earplug
Handla allt 3M Produkter
:
Color: Yellow, Orange
Beskrivning
- Earplug with a orange fitting ring so its easy to when the earplug is correctly fitted
- When the orange color is no longer visible, the best fit is achieved
- Smaller in diameter and length to fit smaller ear canals or those looking for slightly lower attenuation
- The Earplugs are moisture resistant and compatible with other PPE
Specifikationer
Specifikationer
| Färg | Yellow, Orange |
| Par per paket | 200 Pairs/Box |
| Noise Reduction Rating (NRR) | 33 dB |
| Med sladd | Uncorded |
| Produkttyp | Earplugs |
| Dielektrisk | No |