Läs mer
(E)-Alpha-(Methoxyimino)-2-methylbenzeneacetic Acid Methyl Ester, TRC
Produktkod. 30480878
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30480878 250 mg
1 options
Produktkod. 30480878 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM313240250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-alpha-(Methoxyimino)-2-methylbenzeneacetic Acid Methyl Ester
CAS 120974-97-2
Molekylformel C11H13NO3
Kvantitet 250 mg
Synonym (E)-Methyl 2-Methylphenylglyoxylate O-Methyloxime
InChI formel InChI=1S/C11H13NO3/c1-8-6-4-5-7-9(8)10(12-15-3)11(13)14-2/h4-7H,1-3H3/b12-10+
LEDER O=C(OC)/C(C1=CC=CC=C1C)=N/OC
IUPAC-namn methyl (E)-2-(methoxyimino)-2-(o-tolyl)acetate
Molekylvikt (g/mol) 207.23
Formel vikt 207.09