Läs mer
(E)-Cefixime (Cefixime EP Impurity D), TRC
Produktkod. 30363344
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30363344 1 mg
1 options
Produktkod. 30363344 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2428201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cefixime EP Impurity D
CAS 97164-56-2
Molekylformel C16 H15 N5 O7 S2
Kvantitet 1 mg
Synonym Cefixime Imp. D (EP), Cefixime (E)-Isomer, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2E)-2-(2-amino-4-thiazolyl)-2-[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)[(carboxymethoxy)imino]acetyl]amino]-3-ethenyl-8-oxo-, [6R-[6α,7β(E)]]- (9CI), (6R,7R)-7-[((2E)-2-(2-Amino-4-thiazolyl)-2-[(carboxymethoxy)imino]acetyl)amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
InChI formel InChI=1S/C16H15N5O7S2/c1-2-6-4-29-14-10(13(25)21(14)11(6)15(26)27)19-12(24)9(20-28-3-8(22)23)7-5-30-16(17)18-7/h2,5,10,14H,1,3-4H2,(H2,17,18)(H,19,24)(H,22,23)(H,26,27)/b20-9+/t10-,14-/m1/s1
LEDER Nc1nc(cs1)\C(=N/OCC(=O)O)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)C=C
IUPAC-namn (6R,7R)-7-[[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molekylvikt (g/mol) 453.4496
Formel vikt 453.0413