Läs mer
IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion
Handla allt IBA Lifesciences Produkter
Kvantitet:
5 μg
Förpackningsstorlek:
5µg
Beskrivning
- The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
- The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
- The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
- Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Specifikationer
Specifikationer
| Inducerande medel | Inducible with anhydrotetracycline |
| Markör Gene | Ampicillin |
| Promotor | Tet |
| Produkttyp | Vector |
| Innehåll och lagring | Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage |
| Protein Tag | His Tag (6x) |
| Kvantitet | 5 μg |
| Vektor | pASG-IBA33 |