Läs mer
(E)-Daporinad, MedChemExpress
Produktkod. 16474037
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16474037 50 mg
16430588 5 mg
16418128 10 mg
16417886 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16474037 Leverantör Medchem Express Leverantörsnummer HY50876
MedChemExpress (E)-Daporinad (FK866) is an effective inhibitor of nicotinamide phosphoribosyltransferase (NMPRTase; Nampt) with an IC50 of 0.09 nM.
(E)-Daporinad (FK866) is an effective inhibitor of nicotinamide phosphoribosyltransferase (NMPRTase
Specifikationer
Kemiskt namn eller material (E)-Daporinad
Fysisk form Solid
CAS 658084-64-1
Färg White
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C24H29N3O2
För användning med (applikation) Cancer-Kinase/protease
Synonym FK866 APO866
Löslighetsinformation DMSO : ≥ 50 mg/mL (127.71 mM) ∣H2O : 1 mg/mL (2.55 mM; Need ultrasonic)
LEDER O=C(C1=CC=CC=C1)N(CC2)CCC2CCCCNC(/C=C/C3=CC=CN=C3)=O
Molekylvikt (g/mol) 391.51
Formel vikt 391.51
Procent renhet 99.03%