Läs mer
(E)-Dichlorodenafil, TRC
Produktkod. 30370309
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30370309 5 mg
1 options
Produktkod. 30370309 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4340535MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-Dichlorodenafil
CAS 1469486-82-5
Molekylformel C19H20Cl2N4O2
Kvantitet 5 mg
Synonym trans-Dichlorodenafil, 5-[5-[(1E)-1,2-Dichloroethenyl]-2-ethoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one
InChI formel InChI=1S/C19H20Cl2N4O2/c1-4-6-14-16-17(25(3)24-14)19(26)23-18(22-16)12-9-11(13(21)10-20)7-8-15(12)27-5-2/h7-10H,4-6H2,1-3H3,(H,22,23,26)/b13-10+
LEDER O[C@H]1[C@@H](O)[C@H](N2C(NC(C=C2)=O)=S)O[C@@H]1COP(CP(O[Na])(O[Na])=O)(O[Na])=O
Molekylvikt (g/mol) 407.29