Läs mer
(E,E)-2,6-Dimethyl-2,6-octadienedial, TRC
Produktkod. 30375529
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375529 250 mg
30383228 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30375529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD476180250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80054-40-6
Molekylformel C10H14O2
Molekylvikt (g/mol) 166.22
Synonym (2E,6E)-2,6-Dimethyl-2,6-octadienedial
IUPAC-namn (2E,6E)-2,6-dimethylocta-2,6-dienedial
LEDER O=C/C(C)=C/CC/C(C)=C/C=O
Specifikationer
Kemiskt namn eller material (E,E)-2,6-Dimethyl-2,6-octadienedial
Kvantitet 250 mg
InChI formel InChI=1S/C10H14O2/c1-9(6-7-11)4-3-5-10(2)8-12/h5-8H,3-4H2,1-2H3/b9-6+,10-5+
Formel vikt 166.0994