Läs mer
(E,E,E)-2-Acetyl-5,9,13,17-tetramethyl-4,8,12,16-octadecatetraenoic Acid Ethyl Ester, TRC
Produktkod. 30318253
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30318253 50 mg
1 options
Produktkod. 30318253 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA18862050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E,E,E)-2-Acetyl-5,9,13,17-tetramethyl-4,8,12,16-octadecatetraenoic Acid Ethyl Ester
CAS 71668-81-0
Molekylformel C26H42O3
Kvantitet 50 mg
Synonym ethyl (4E,8E,12E)-2-acetyl-5,9,13,17-tetramethyloctadeca-4,8,12,16-tetraenoate 4,8,12,16-Octadecatetraenoic acid, 2-acetyl-5,9,13,17-tetramethyl-, ethyl ester, (E,E,E)- (9CI)
InChI formel InChI=1S/C26H42O3/c1-8-29-26(28)25(24(7)27)19-18-23(6)17-11-16-22(5)15-10-14-21(4)13-9-12-20(2)3/h12,14,16,18,25H,8-11,13,15,17,19H2,1-7H3/b21-14+,22-16+,23-18+
LEDER C/C(C)=C\CC/C(C)=C/CC/C(C)=C/CC/C(C)=C/CC(C(C)=O)C(OCC)=O
IUPAC-namn ethyl (4E,8E,12E)-2-acetyl-5,9,13,17-tetramethyloctadeca-4,8,12,16-tetraenoate
Molekylvikt (g/mol) 402.61
Formel vikt 402.313