Läs mer
(E)-Fluvoxamine Maleate, TRC
Produktkod. 30009059
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009059 25 mg
17984385 10 mg
17940385 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF60350025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61718-82-9
Molekylformel C15 H21 F3 N2 O2 . C4 H4 O4
Molekylvikt (g/mol) 434.41
Synonym Fluvoxamine maleate, 2-[[[(1E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxy]ethanamine (Z)-butenedioate, 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (1E)-, (2Z)-2-butenedioate (1:1) (9CI, ACI), 1-Pentanone, 5-methoxy-1-[4-(trifluoromethyl)phenyl]-, O-(2-aminoethyl)oxime, (E)-, (Z)-2-butenedioate (1:1) (ZCI), Avoksin, Avoxin, Depromel, DU 23000, Dumirox, Faverin, Faverin 50, Faverine, Fevarin, Floxyfral, Luvox, Maveral, MK 264, NSC 309469
IUPAC-namn (Z)-but-2-enedioic acid;2-[(E)-[5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene]amino]oxyethanamine
LEDER COCCCC\C(=N/OCCN)\c1ccc(cc1)C(F)(F)F.OC(=O)\C=C/C(=O)O
Specifikationer
Kemiskt namn eller material Fluvoxamine maleate
Kvantitet 25 mg
InChI formel InChI=1S/C15H21F3N2O2.C4H4O4/c1-21-10-3-2-4-14(20-22-11-9-19)12-5-7-13(8-6-12)15(16,17)18;5-3(6)1-2-4(7)8/h5-8H,2-4,9-11,19H2,1H3;1-2H,(H,5,6)(H,7,8)/b20-14+;2-1-
Formel vikt 434.1665