Läs mer
(E)-Guggulsterone, TRC
Produktkod. 30240983
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240983 100 mg
30240749 50 mg
30240494 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240983 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG852005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 39025-24-6
Molekylformel C21 H28 O2
Molekylvikt (g/mol) 312.45
Synonym Pregna-4,17(20)-diene-3,16-dione, (17E)-, (-)-(E)-Guggulsterone, (17E)-Pregna-4,17(20)-diene-3,16-dione, 4,17(20)-(trans)-Pregnadiene-3,16-dione, E-Guggulsterone, Guggulsterone E, trans-Guggulsterone
IUPAC-namn (8R,9S,10R,13S,14S,17E)-17-ethylidene-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthrene-3,16-dione
LEDER C\C=C/1\C(=O)C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Guggulsterone E
Kvantitet 100 mg
InChI formel InChI=1S/C21H28O2/c1-4-16-19(23)12-18-15-6-5-13-11-14(22)7-9-20(13,2)17(15)8-10-21(16,18)3/h4,11,15,17-18H,5-10,12H2,1-3H3/b16-4-/t15-,17+,18+,20+,21-/m1/s1
Formel vikt 312.2089