Läs mer
(E)-Hex-3-en-1-yl Acetate, TRC
Produktkod. 30136019
Change view
Kvantitet:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136019 500 mg
30006757 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136019 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH293640500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3681-82-1
Molekylformel C8H14O2
Molekylvikt (g/mol) 142.2
Synonym (3E)-Hexenyl Acetate, (E)-3-Hexen-1-ol Acetate, (E)-3-Hexen-1-yl Acetate, (E)-3-Hexenyl Acetate, Acetic Acid trans-3-Hexenyl Ester, E-3-Hexen-1-yl Acetate, trans-3-Hexen-1-ol Acetate, trans-3-Hexen-1-yl Acetate, trans-3-Hexenyl Acetate;
IUPAC-namn [(E)-hex-3-enyl] acetate
LEDER CC\C=C\CCOC(=O)C
Specifikationer
Kemiskt namn eller material (E)-Hex-3-en-1-yl Acetate
Kvantitet 500 mg
InChI formel InChI=1S/C8H14O2/c1-3-4-5-6-7-10-8(2)9/h4-5H,3,6-7H2,1-2H3/b5-4+
Formel vikt 142.0994