Läs mer
(E)-Methyl 3-(Methoxy(phenyl)methylene)-2-oxoindoline-6-carboxylate, TRC
Produktkod. 30479410
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30479410 500 mg
30475327 250 mg
30475885 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30479410 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM322025500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1168150-46-6
Molekylformel C18 H15 N O4
Molekylvikt (g/mol) 309.316
Synonym 1H-Indole-6-carboxylic acid, 2,3-dihydro-3-(methoxyphenylmethylene)-2-oxo-, methyl ester, (3E)- (ACI), Methyl (3E)-2,3-dihydro-3-(methoxyphenylmethylene)-2-oxo-1H-indole-6-carboxylate (ACI)
IUPAC-namn methyl (3E)-3-[methoxy(phenyl)methylidene]-2-oxo-1H-indole-6-carboxylate
LEDER COC(=O)c1ccc2\C(=C(/OC)\c3ccccc3)\C(=O)Nc2c1
Specifikationer
Kemiskt namn eller material (E)-Methyl 3-(Methoxy(phenyl)methylene)-2-oxoindoline-6-carboxylate
Kvantitet 500 mg
InChI formel InChI=1S/C18H15NO4/c1-22-16(11-6-4-3-5-7-11)15-13-9-8-12(18(21)23-2)10-14(13)19-17(15)20/h3-10H,1-2H3,(H,19,20)/b16-15+
Formel vikt 309.1