Läs mer
(E)-N-(3-Methylaminopropyl)-2-thiopheneacrylamide Hydrochloride, TRC
Produktkod. 30481153
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481153 25 mg
30473839 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30481153 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28697025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1449108-90-0
Molekylformel C11H17ClN2OS
Molekylvikt (g/mol) 260.78
Synonym (2E)-N-[3-(Methylamino)propyl]-3-(thiophen-2-yl)prop-2-enamide Hydrochloride, (E)-N-[3-(Methylamino)propyl]-3-(2-thienyl)-2-propenamide Hydrochloride
LEDER N#Cc1ccc(N)cc1
Specifikationer
Kemiskt namn eller material (E)-N-(3-Methylaminopropyl)-2-thiopheneacrylamide Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C11H16N2OS.ClH/c1-12-7-3-8-13-11(14)6-5-10-4-2-9-15-10;/h2,4-6,9,12H,3,7-8H2,1H3,(H,13,14);1H/b6-5+;