Läs mer
(E)-Neochlorogenic Acid, TRC
Produktkod. 17932085
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17932085 10 mg
17942955 50 mg
17940755 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17932085 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN38990510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 906-33-2
Molekylformel C16 H18 O9
Molekylvikt (g/mol) 354.31
Synonym Cyclohexanecarboxylic acid, 3-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,4,5-trihydroxy-, (1R,3R,4S,5R)-, Cyclohexanecarboxylic acid, 3-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxy-, (1R,3R,4S,5R)- (9CI), Cyclohexanecarboxylic acid, 3-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxy-, [1R-[1alpha,3alpha(E),4alpha,5beta]]-, Neochlorogenic acid (6CI,8CI), (1R,3R,4S,5R)-3-[[(2E)-3-(3,4-Dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid, (E)-Neochlorogenic acid, 3-O-(E)-Caffeoylquinic acid, 5-(E)-Caffeoylquinic acid, 5-Caffeoylquinic acid, 5-O-Caffeolyquinic acid, 5-O-Caffeoylquinic acid, trans-5-O-Caffeoylquinic acid
IUPAC-namn (1R,3R,4S,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid
LEDER O[C@@H]1C[C@@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@H]1O)C(=O)O
Specifikationer
Kemiskt namn eller material (E)-Neochlorogenic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14+,16-/m1/s1
Formel vikt 354.0951
Procent renhet >95%