Läs mer
(E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-(4-hydroxyphenyl)-2-phenyl-1-butene, TRC
Produktkod. 30352727
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352727 100 mg
30352271 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30352727 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC365785100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119757-57-2
Molekylformel C24H23ClO2
Molekylvikt (g/mol) 378.89
Synonym 4-[1-[4-(2-Chloroethoxy)phenyl]-2-phenyl-1-butenyl]-phenol
IUPAC-namn 4-[(E)-1-[4-(2-chloroethoxy)phenyl]-2-phenylbut-1-enyl]phenol
LEDER OC1=CC=C(/C(C2=CC=C(OCCCl)C=C2)=C(CC)/C3=CC=CC=C3)C=C1
Specifikationer
Kemiskt namn eller material (E/Z)-1-[4-(2-Chloroethoxy)phenyl]-1-(4-hydroxyphenyl)-2-phenyl-1-butene
Kvantitet 100 mg
InChI formel InChI=1S/C24H23ClO2/c1-2-23(18-6-4-3-5-7-18)24(19-8-12-21(26)13-9-19)20-10-14-22(15-11-20)27-17-16-25/h3-15,26H,2,16-17H2,1H3/b24-23+
Formel vikt 378.1387