Läs mer
(E/Z)-4-Hydroxy Tamoxifen, TRC
Produktkod. 30133938
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133938 25 mg
30012171 5 mg
30005345 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133938 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95470525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68392-35-8
Molekylformel C26H29NO2
Molekylvikt (g/mol) 387.51
Synonym 4-[1-[4-[2-(Dimethylamino)ethoxy]phenyl]-2-phenyl-1-butenyl]phenol, (E/Z)-4-Hydroxytamoxifen, 4-Hydroxytamoxifen, 4-Hydroxy-(E/Z)-tamoxifen, Afimoxifene, TamoGel;
LEDER COC(=O)[C@H](CCl)NC(C)=O
Specifikationer
Kemiskt namn eller material (E/Z)-4-Hydroxy Tamoxifen
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C26H29NO2/c1-4-25(20-8-6-5-7-9-20)26(21-10-14-23(28)15-11-21)22-12-16-24(17-13-22)29-19-18-27(2)3/h5-17,28H,4,18-19H2,1-3H3/b26-25-
Formel vikt 387.2198
Procent renhet >95%