Läs mer
(E/Z)-N,N-Didesmethyl-4-hydroxy Tamoxifen, TRC
Produktkod. 30137282
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137282 5 mg
30012835 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137282 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4411205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1217237-98-3
Molekylformel C24H25NO2
Molekylvikt (g/mol) 359.46
Synonym 4-[1-[4-(2-Aminoethoxy)phenyl]-2-phenyl-1-buten-1-yl]phenol;
LEDER CCC(=C(C1=CC=C(C=C1)O)C2=CC=C(C=C2)OCCN)C3=CC=CC=C3
Specifikationer
Kemiskt namn eller material (E/Z)-N,N-Didesmethyl-4-hydroxy Tamoxifen
Kvantitet 5 mg
InChI formel InChI=1S/C24H25NO2/c1-2-23(18-6-4-3-5-7-18)24(19-8-12-21(26)13-9-19)20-10-14-22(15-11-20)27-17-16-25/h3-15,26H,2,16-17,25H2,1H3/b24-23-
Formel vikt 359.1885