Läs mer
Thermo Scientific™ E1-ClipTip™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes
Produktkod. 15236283
Change view
Volym (metrisk):
0,5 till 12,5 μl
1 till 30 μl
10 till 300 μl
15 till 1250 μl
2 till 125 μl
Antal kanaler:
6
8
12
Kompatibla tips:
ClipTip 1 250
ClipTip 200
ClipTip 300
ClipTip 384 12,5
Klipptips 384 125
Klipptips 384 30
Förpackningsstorlek:
Varje
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Volym (metrisk) Antal kanaler Kompatibla tips Förpackningsstorlek
15236283 0,5 till 12,5 μl 8 ClipTip 384 12,5 Varje
15206283 2 till 125 μl 8 ClipTip 200 Varje
15296273 15 till 1250 μl 6 ClipTip 1 250 Varje
15216283 10 till 300 μl 8 ClipTip 300 Varje
15246283 1 till 30 μl 8 Klipptips 384 30 Varje
15256283 2 till 125 μl 8 Klipptips 384 125 Varje
15266283 0,5 till 12,5 μl 12 ClipTip 384 12,5 Varje
15276283 1 till 30 μl 12 Klipptips 384 30 -
15286283 2 till 125 μl 12 Klipptips 384 125 Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Produktkod. 15236283 Leverantör Thermo Scientific™ Leverantörsnummer 4672010
 Begränsad artikel
Beräknad leverans: 07-12-2026
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with adjustable tip spacing.
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip™ Equalizer Pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format. Feel the difference.
Innovative ClipTip interlocking tip interface technology†
  • Tip is locked and sealed in place until ejected— no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch— eliminates the frustration of banging tips on the pipette
An ideal choice for sample transfers
Reduce repetition and save time between labware formats with adjustable tip spacing in Equalizer models
Unified operation for multiple users and environments
  • Personalized user interface and password protection ensure consistent operation among multiple users.
  • Electronic operation to help you enjoy pipetting again
  • Designed for optimum comfort and ease of use, the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
Two operating options ensure optimal function for every application
Choose a step based Matrix or Presets with settings for your most common applications
MyPipette™ - One click access for more efficient and unified pipetting
Customize your main menu view by organizing icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
Store up to 20 programs
Tired of continuous editing? The Programs function allows you to store 20 your most common Matrix or Presets protocols with a specific name.
A pipette for every application
Choose from single channels, 8-16 channel multichannels, or 6- 12-channel adjustable tip spacing Equalizer multichannel pipettes within the volume range of 0.5-1250μL.
Track your pipette
Service and calibration trackers remind you when the pipette needs to be sent to serviced
Liquid specific calibrations
Higher quality results with liquid specific calibrations
Li-Ion battery
Long lasting performance and fast recharging.
Flexible re-charging
Use either charging stand or a plug, you decide
2-year warranty
With convenient web registration
Compatible with Thermo Scientific ClipTip pipette tips.
† E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifikationer
Noggrannhet ±2.5 to 12.0%
Kompatibla tips ClipTip 384 12,5
Beskrivning E1-ClipTip Equalizer; Format: 384
Justerbart spetsavstånd Yes
Färg Pink
För användning med ClipTip 384 12.5
Modell E1-ClipTip
Antal kanaler 8
Produktlinje E1-ClipTip
Programmerbarhet Yes
Typ Electronic Pipette
Volym (metrisk) 0,5 till 12,5 μl
Ökar 0.01μL
Precision ≤1.6 to 12.0%