missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ E2F3 (Human) Recombinant Protein
Human E2F3 full-length ORF ( AAH16847, 1 a.a. - 133 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001871-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MQSGGGVKTDDTSTLNSLCGYAWVYVWEEKQRCRLSSFFSSSASIPGLLPSHTLDLVQNVGVVLDEALGWGRERELCVKCLLEMHCGVFSCMGNHLCQAFPHFPYLSHLVSCLCFQLCVILFASCTKLIFSKV
Specifications
AAH16847 | |
E2F transcription factor 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E2F3 | |
GST |
1871 | |
Wheat germ expression system | |
25 μg | |
DKFZp686C18211, E2F-3, KIAA0075, MGC104598 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |