Learn More
EAF1 Monoclonal Antibody (1G2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524498
Additional Details : Weight : 0.01000kg
Description
Peptide Sequence: MNGTANPLLD REEHCLRLGE SFEKRPRASF HTIRYDFKPA SIDTSCEGEL QVGKGDEVTI TLPHIPGSTP PMTVFKGNKR PYQKDCVLII NHDTGEFVLE KLSSSIQVKK TRAEGSSKIQ ARMEQQPTRP PQTSQPPPPP PPMPFRAPTK PPVGPKTSPL KDNPSPEPQL DDIKRELRAE VDIIEQMSSS SGSSSSDSES SSGSDDDSSS SGGEDNGPAS PPQPSHQQPY NSRPAVANGT SRPQGSNQLM NALRNDLQLS ESGSDSDD
EAF1 Monoclonal antibody specifically detects EAF1 in Human samples. It is validated for ELISA, Western Blot.Specifications
EAF1 | |
Monoclonal | |
Unconjugated | |
EAF1 | |
4933403C17Rik; AI666536; EAF1; ELL (eleven nineteen lysine-rich leukemia gene)-associated factor 1; ELL associated factor 1; ELL-associated factor 1 | |
Mouse | |
Protein A | |
RUO | |
85403 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1G2 | |
PBS with no preservative; pH 7.4 | |
Q96JC9 | |
EAF1 | |
EAF1 (AAH41329, 1 a.a. ∽ 268 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |