Learn More
STEMCELL Technologies EasyEights™ EasySep™ Magnet
Product Code. 16645645
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Cell Isolation
Unit Size:
Each
Product Code. For Use With (Application) Unit Size
16645645 Cell Isolation Each
1 options
Product Code. 16645645 Supplier STEMCELL Technologies Supplier No. 18103
Beräknad leverans: 18-11-2026
Immunomagnetic column-free magnet
  • The EasyEights™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when simultaneously processing multiple samples, up to eight samples on each side, a total of 16 samples
  • The EasyEights™ EasySep™ Magnet has a large sample range, capable of processing small samples, starting at 0.1 x 10^8 cells, to larger samples, up to 8.5 x 10^8 cells
  • The EasyEights™ EasySep™ Magnet is designed to hold up to 8 standard 5 mL (12 x 75 mm) round-bottom tubes on one side, and 8 standard 14 mL (17 x 95 mm) round-bottom or 15 mL conical tubes on the other side
Specifications
For Use With (Application) Cell Isolation
Quantity Simultaneously processes up to 8 samples per side, total of 16 samples