Learn More
Trajan Scientific and Medical™ EasyLOK Fitting
Composed of knurled stainless steel nut and a double-ended PEEK ferrule for a leak-free connection.
Brand:  Trajan Scientific and Medical™ 206102
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15239203
  • 580.00 SEK / Pack of 2
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
SGE GLT Columns
5000 psi
EasyLok Fitting
EasyLOK Fitting 2 fittings, 2 ferrules
10-32