Learn More
Trajan Scientific and Medical™ EasyLOK Fitting
Composed of knurled stainless steel nut and a double-ended PEEK ferrule for a leak-free connection.
Brand:  Trajan Scientific and Medical™ 206160
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15249203
  • 740.00 SEK / Pack of 10
Estimated Shipment: 05-07-2024
to see stock.
Add to basket
Description
Description
  • Leak free connections
  • Replacement parts such as nuts and ferrules
Specifications
Specifications
SGE GLT Columns
5000 psi
PEEK Double Ended Ferrules
PEEK Double Ended Ferrules
10-32