Learn More
Trajan Scientific and Medical™ EasyLOK Fitting
Composed of knurled stainless steel nut and a double-ended PEEK ferrule for a leak-free connection.
Brand: Trajan Scientific and Medical™ 206102
Additional Details : Weight : 0.01000kg
Description
- Leak free connections
- Replacement parts such as nuts and ferrules
Specifications
SGE GLT Columns | |
5000 psi | |
EasyLok Fitting |
EasyLOK Fitting 2 fittings, 2 ferrules | |
10-32 |