missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
The protein encoded by this gene was first identified by its binding to the APC protein which is often mutated in familial and sporadic forms of colorectal cancer. This protein localizes to microtubules, especially the growing ends, in interphase cells. During mitosis, the protein is associated with the centrosomes and spindle microtubules. The protein also associates with components of the dynactin complex and the intermediate chain of cytoplasmic dynein. Because of these associations, it is thought that this protein is involved in the regulation of microtubule structures and chromosome stability. This gene is a member of the RP/EB family.
Specifikationer
Specifikationer
| Antigen | EB1 |
| Användningsområden | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.25 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gen | MAPRE1 |
| Genaccessionsnr. | Q15691, Q61166 |
| Gene Alias | APC binding protein EB1, EB1, End binding protein 1, MAPRE1 |
| Gensymboler | Mapre1 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?