missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ EBF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530985
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'.
Specifikationer
| EBF3 | |
| Polyclonal | |
| Unconjugated | |
| Ebf3 | |
| 3110018A08Rik; COE3; early B cell factor 3; early B-cell factor 3; EBF transcription factor 3; EBF3; EBF-3; mKIAA4201; O/E-2; OE-2; olf-1/EBF-like 2; Transcription factor COE3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 253738 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.34 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9H4W6 | |
| Ebf3 | |
| Recombinant protein encompassing a sequence within the center region of human EBF3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Spot an opportunity for improvement?Share a Content Correction