Läs mer
EBI-2511, MedChemExpress
Produktkod. 16473086
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16473086 25 mg 25mg
16464448 10 mg -
16415336 100 mg 100mg
16407177 5 mg -
16400767 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16473086 Leverantör Medchem Express Leverantörsnummer HY111418
MedChemExpress EBI-2511 is a highly potent and orally active EZH2 inhibitor, with an IC50 of 6 nM in Pfeffiera cell lines, respectively.
EBI-2511 is a highly potent and orally active EZH2 inhibitor, with an IC50 of 6 nM in Pfeffiera cell lines, respectively.
Specifikationer
Kemiskt namn eller material EBI-2511
Fysisk form Solid
CAS 2098546-05-3
Färg White
Kvantitet 25 mg
Anteckningar om renhetsgrad Research
Molekylformel C34H48N4O4
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 5 mg/mL (8.67 mM; Need ultrasonic)
LEDER CCC1=C(C(NCC2=C(C)C=C(C)NC2=O)=O)C(C=C(C3CCN(C(C)C)CC3)O4)=C4C=C1N(C5CCOCC5)CC
Molekylvikt (g/mol) 576.77
Formel vikt 576.77
Procent renhet 99.41%