Learn More
EBLN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566501
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: QALLSVGVSKR SNTVVGNENE ERGTPYASRF KDMPNFIALE KSSVLRHCCD LLIGIAAGSS DKICTSS Highest antigen sequence identity to the following orthologs - mouse 29%, rat 29%.
May act as an RNA-binding protein. The C-terminal region is highly homologous to the bornavirus nucleocapsid N protein that binds viral RNA and oligomerizes. The viral protein also possesses a nuclear import and a nuclear export signal. These 2 signals seem absent in EBLN-2 supporting an unrelated function in Human.Specifications
EBLN2 | |
Polyclonal | |
Unconjugated | |
EBLN2 | |
EBLN-1; EBLN2; EBLN-2; endogenous Borna-like N element-1; endogenous Borna-like N element-2; Endogenous Bornavirus-like nucleoprotein 2; GK006; ikappaBalpha; I-kappa-B-alpha; Ikba; ikB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; NF-kappa-B inhibitor alpha; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; RL/IF-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55096 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P2I7 | |
EBLN2 | |
Recombinant Human EBLN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only