missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECD Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10192-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
ERp72 is an endoplasmic reticulum luminal protein that is both a stress protein and a member of the protein disulfide isomerase family of proteins. Sequence analysis of ERp72 shows that it shares sequence identity with PDI (protein disulphide isomerase) at three discrete regions, as well as containing the ER retention signal KEEL at its C-terminus. ERp72 is shown to have three copies of sequence believed to be CGHC-containing active sites, which are also found in PDI. These CGHC sites are critical sites that are involved in the protein disulphide isomerization process. These results suggest that ERp72 is a newly described member of the family of CGHC-containing proteins that functions as a molecular chaperone.Specifications
ECD | |
Polyclonal | |
Unconjugated | |
ECD | |
ECD, GCR2, HSGT1, Protein ecdysoneless homolog, Protein SGT1, SGT1, Suppressor of GCR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11319, 70601 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95905, Q9CS74 | |
Ecd | |
ECD Fusion Protein Ag0245 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |