Learn More
ECD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552298
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DEDFECLDSD DDLDFETHEP GEEASLKGTL DNLKSYMAQM DQELAHTCIS KSFTTRNQVE PVSQTTDNNS DEEDSGTGES VMAPVDVDLN LVSNILESYS SQAGLAGPAS NLLQSMGVQL PDNTDHRPTS KP Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 69%.
ECD Polyclonal antibody specifically detects ECD in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
ECD | |
Polyclonal | |
Unconjugated | |
ECD | |
5730461K03Rik; Ecd; ecdysoneless cell cycle regulator; ecdysoneless homolog; ecdysoneless homolog (Drosophila); GCR2; HSGT1; Human suppressor of GCR two; protein ecdysoneless homolog; protein SGT1; protein SGT1 homolog; SGT1; SGT1 protein; suppressor of GCR2; suppressor of S. cerevisiae gcr2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11319 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.08 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95905 | |
ECD | |
Recombinant protein corresponding to Human ECD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only