missing translation for 'onlineSavingsMsg'
Learn More

Labcon Eclipse™ 300 μL Pipette Tips

Fits Most Popular 300 μL Pipettors

Brand:  Labcon 1039-965-006-9

Additional Details : Weight : 14.06140kg

 View more versions of this product

Product Code. 16623912

  • 697.00 SEK / Pack of 960
Estimated Shipment: 24-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Pipette Tip
  • Fits Most Popular 300 μL Pipettors
  • Beveled Point
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Autoclavable
  • Made in USA
Specifications

Specifications

Yes
Polypropylene USP Class VI resin
300 μL
Sterile
Autoclavable
52.1 mm
ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
E-Beam
Beveled
96 Tips/Rack
No
Eclipse™
Non-Filtered
Clear
2.05 in.
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity
Liquid Handing
Fits Most Popular 300 µL Pipettors
96 Tips/Rack; 10 Racks/Pk.; 10 Pk./Cs.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers