Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 65 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
755.00 SEK - 1090.00 SEK
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 65 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408200
|
Tenak™
TE15200 |
Microtiterplates (9) |
995.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
12428200
|
Tenak™
TE15204 |
Microtiterplates (11) |
1090.00 SEK
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
12458190
|
Tenak™
TE15190 |
Microtiterplates (4) |
755.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
12468190
|
Tenak™
TE15192 |
Microtiterplates (5) |
795.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
12478190
|
Tenak™
TE15194 |
Microtiterplates (6) |
850.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
12488190
|
Tenak™
TE15196 |
Microtiterplates (7) |
890.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
12498190
|
Tenak™
TE15198 |
Microtiterplates (8) |
935.00 SEK
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Chest Frseezers | |
65 mm |