missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ ECO AluRack for Chest Freezers, 100 mm Box Height, with Boxes
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
950.00 SEK - 1290.00 SEK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Description | Seawater resistant |
Material | Aluminum |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 100 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12418150
|
Tenak™
TE11560B |
Boxes (3) |
950.00 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
12438150
|
Tenak™
TE11562B |
Boxes (4) |
1050.00 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
12458150
|
Tenak™
TE11564B |
Boxes (5) |
1180.00 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
12478150
|
Tenak™
TE11566B |
Boxes (6) |
1290.00 SEK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Aluminum | |
100 mm (Box) |
Seawater resistant | |
Chest Frseezers |