Learn More
Tenak™ ECO AluRack for Upright Freezers, 32 mm Box Height, with Boxes
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
1520.00 SEK - 2725.00 SEK
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Description | Seawater resistant |
Material | Aluminum |
Height (Metric) | 32 mm (Box) |
Product Code | Brand | Holds | Array | Dimensions (L x W x H) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Dimensions (L x W x H) | Price | Quantity & Availability | |||
12402480
|
Tenak™
TE11122B |
Boxes (18) | 6 x 3 | 425 x 139 x 216 mm |
1675.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12402490
|
Tenak™
TE11134B |
Boxes (28) | 7 x 4 | 564 x 139 x 252 mm |
2175.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12402500
|
Tenak™
TE11152B |
Boxes (30) | 6 x 5 | 693 x 139 x 216 mm |
2255.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12422480
|
Tenak™
TE11124B |
Boxes (21) | 7 x 3 | 425 x 139 x 252 mm |
1835.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12422490
|
Tenak™
TE11136B |
Boxes (32) | 8 x 4 | 564 x 139 x 288 mm |
2355.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12422500
|
Tenak™
TE11154B |
Boxes (35) | 7 x 5 | 693 x 139 x 252 mm |
2485.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12442480
|
Tenak™
TE11126B |
Boxes (24) | 6 x 4 | 425 x 139 x 288 mm |
1985.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12442490
|
Tenak™
TE11138B |
Boxes (36) | 9 x 4 | 564 x 139 x 324 mm |
2520.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12442500
|
Tenak™
TE11156B |
Boxes (40) | 8 x 5 | 693 x 139 x 288 mm |
2725.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12462480
|
Tenak™
TE11130B |
Boxes (20) | 5 x 4 | 564 x 139 x 180 mm |
1735.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12462490
|
Tenak™
TE11140B |
Boxes (40) | 10 x 4 | 564 x 139 x 360 mm |
2725.00 SEK
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
12482470
|
Tenak™
TE11120B |
Boxes (15) | 5 x 3 | 425 x 139 x 180 mm |
1520.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12482480
|
Tenak™
TE11132B |
Boxes (24) | 6 x 4 | 564 x 139 x 216 mm |
1930.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
12482490
|
Tenak™
TE11150B |
Boxes (25) | 5 x 5 | 683 x 139 x 180 mm |
2025.00 SEK
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Includes standard white cardboard boxes with 9 x 9 cell dividers
- Holds is expressed by Height x Dept
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Seawater resistant | |
Aluminum | |
32 mm (Box) |