Läs mer
Econazole EP Impurity B Hydrochloride, TRC
Produktkod. 30370328
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30370328 100 mg
1 options
Produktkod. 30370328 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE344510100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Econazole EP Impurity B Hydrochloride
CAS 2334059-11-7
Molekylformel C15H14Cl3NO . HCl
Kvantitet 100 mg
Synonym 2-((4-chlorobenzyl)oxy)-2-(2,4-dichlorophenyl)ethan-1-amine, hydrochloride (1:1)
InChI formel InChI=1S/C15H14Cl3NO/c16-11-3-1-10(2-4-11)9-20-15(8-19)13-6-5-12(17)7-14(13)18/h1-7,15H,8-9,19H2
LEDER ClC1=CC=C(COC(CN)C2=CC=C(C=C2Cl)Cl)C=C1.Cl
Molekylvikt (g/mol) 330.6