Learn More
Safety First Aid Economy Workplace First Aid Kit, HSE Compliant (21 to 50 Persons)
Product Code. 31759428
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
Product Code. Sterility Unit Size
31759428 Sterile Each
1 options
Product Code. 31759428 Supplier Safety First Aid Supplier No. K50AECON
 Restricted Item
Beräknad leverans: 27-11-2026
The kit contains all the sterile first aid dressings, plasters, wipes and gloves that are recommended by the HSE. It is fully compliant and up to date with all the HSE regulations and supplied in a lightweight, easily accessible green box with locking clips.
  • Cases can be made even more visible and accessible with our wall bracket (sold separately).
  • Once used, this case can be retained and refilled in the future with our HSE Workplace First Aid Kit Refill 21-50 Persons.
  • The HSE Economy Workplace First Aid Kit (21-50 Person) Contains:
    • 1x First Aid Guidance Leaflet
    • 60x HypaPlast Washproof Plastes
    • 6x HypaCover Eye Dressings
    • 6x HypaBand Triangular Bandages
    • 12x HypaBand Safety Pins
    • 12x HypaCover First Aid Dressings, 12 x 12 cm
    • 4x HypaCover First Aid Dressings, 18 x 18 cm
    • 20x HypaCover Sterile Moist Wipes, Alcohol-Free
    • 5x HypaTouch Disposable Gloves
Specifications
Product Type First Aid Kit
Material Plastic
Sterility Sterile