missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ EDA Monoclonal Antibody (174806)

Beskrivning
Reconstitute at 0.5 mg/mL in sterile PBS.
EDA is a type II membrane protein that can be cleaved by furin to produce a secreted form. This protein, which belongs to the tumor necrosis factor family, acts as a homotrimer and may be involved in cell-cell signaling during the development of ectodermal organs. Defects in the gene for EDA are a cause of ectodermal dysplasia, anhidrotic, which is also known as X-linked hypohidrotic ectodermal dysplasia.
Specifikationer
Specifikationer
| Antigen | EDA |
| Användningsområden | ELISA |
| Klassificering | Monoclonal |
| Klona | 174806 |
| Koncentration | 0.5 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with trehalose and no preservative |
| Gen | EDA |
| Genaccessionsnr. | Q92838 |
| Gene Alias | ECTD1; ectodermal dysplasia 1, anhidrotic; ectodermal dysplasia protein; Ectodysplasin; Ectodysplasin 1, isoform A2; ectodysplasin A; ectodysplasin-1; Ectodysplasin-A; Ectodysplasin-A, membrane form; Ectodysplasin-A, secreted form; Ed1; ED1-A1; ED1-A2; EDA; EDA 1; EDA protein; EDA protein homolog; EDA1; EDA2; Eda-A1; EDA-A2; HED; HED1; nackt; nkt; ODT1; oligodontia 1; RGD1563178; si:ch73-223d24.5; STHAGX1; Ta; tabby; Tabby protein; TNLG7C; tumor necrosis factor ligand 7C; XHED; XLHED; X-linked anhidroitic ectodermal dysplasia protein |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?